Ciprofloxacin Hydrochlorid Monohydrat ≥98% (durch HPLC)

Lieferant: ENZO LIFE SCIENCES

Synonyme: 1-Cyclopropyl- 6-Fluoro-1,4-dihydro-4-oxo-7-(1-piperazinyl)-3-quinolinecarboxylic acid hydrochloride monohydrate, 1-Cyclopropyl-6-fluoro-4-oxo-7-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride monohydrate, Nanomycopulitine

ALX380288G005
ENZOALX380288G005EA 51.7 EUR
ENZOALX380288G005
Ciprofloxacin Hydrochlorid Monohydrat ≥98% (durch HPLC)
Ciprofloxacin hydrochloride monohydrate

Fluoroquinolone antibacterial agent. Inhibits bacterial DNA gyrase (topoisomerase). Inhibits cell division and causes double-strand breaks in the bacterial chromosome. Broad-spectrum antibiotic effective against Gram-positive, Gram-negative and anaerobic bacteria.


  • Effective against Mycobacterium tuberculosis
  • Potentially anxiogenic


White to light yellow powder


Soluble in water (35 mg/ml)


Verpackung: Shipping temperature: Ambient

Formel: C₁₇H₁₈FN₃O₃·HCl·H₂O
Molecular Weight: 385,82 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00242856
CAS-Nummer: 86393-32-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR