Hygromycin B 100 mg/ml in Wasser (aus Streptomyces hygroscopicus) , 90% (durch HPLC), steril gefiltert
Lieferant: ENZO LIFE SCIENCES
Synonyme:
(3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
Antibiotic. Inhibits growth of prokaryotic and eukaryotic microorganisms and mammalian cells. Inhibits protein synthesis at the translocation step on the 70S ribosome and causes misreading of the mRNA. Penetrates cells that have been permeabilized by virus infection, hence it can act as an effective antiviral agent.
- Protein synthesis inhibitor
Liquid. In deionized water
Formel:
C₂₀H₃₇N₃O₁₃ Molecular Weight: 527,5 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD06795479 CAS-Nummer: 31282-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...