Griess reagent
Lieferant: ENZO LIFE SCIENCES
ALX400004L050
ENZOALX400004L050EA 119 EUR
ENZOALX400004L050
Griess reagent
Griess reagent
Reagent for nitrite detection.
Reagent for the determination of nitrite (NO²⁻) in biological media. Consists of 1 part 0.1% naphthylethylenediamine dihydrochloride in distilled water plus 1 part 1% sulfanilamide (or sulphanilic acid) in 25 - 50% acetic acid.
Soluble in water.