Cytidine-5'-diphosphocholine sodium salt (CDP choline) ≥99%

Lieferant: ENZO LIFE SCIENCES

Synonyme: CDP-Choline, 2-(((((((2R,3S,4R,5R)-5-(4-Amino-2-oxopyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)-N,N,N-trimethylethan-1-aminium, sodium salt(1:1), Citicoline sodium salt, CDP-choline sodium salt

BMLEI1231000
ENZOBMLEI1231000EA 231 EUR
ENZOBMLEI1231000
Cytidine-5'-diphosphocholine sodium salt (CDP choline) ≥99%
Cytidine-5'-diphosphocholine sodium salt (CDP choline)

Completely inhibits PLA2 activation following cryogenic brain injury in the rabbit at 200 mg/kg. Intermediate in phospholipid biosynthesis.


  • Phospholipase inhibitor


White solid


Soluble in water.

Formel: C₁₄H₂₆N₄NaO₁₁P₂
Molecular Weight: 510,31 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00037486
CAS-Nummer: 33818-15-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07f4cc7b533fdb7faa069f14f17fb6f0e8f55212&vid=894449d45a6f1c538e0dbc7b03fc919894dd775a&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR