Bumetanide ≥98%

Lieferant: ENZO LIFE SCIENCES

BMLEI1680005
ENZOBMLEI1680005EA 257 EUR
ENZOBMLEI1680005
Bumetanide ≥98%
Bumetanide

Na+ channel blocker


Bumetanide is a specific inhibitor of Na-K-2Cl cotransport. It binds specifically, saturably and with high affinity to membranes from a large variety of cells. A clinically useful diuretic.

Formel: C₁₇H₂₀N₂O₅S
MDL: MFCD00078949
CAS-Nummer: 28395-03-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=197fcb318baad1aa9d0e7261872089154c4a67ca&vid=280e0d757fe37bf5a4c7b49a222875d9bdd5d3ea&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR