Mu-Phe-hPhe-FMK, Enzo Life Sciences
Lieferant: ENZO LIFE SCIENCES
BMLEI3230001
ENZOBMLEI3230001EA
260
EUR
ENZOBMLEI3230001
Mu-Phe-hPhe-FMK, Enzo Life Sciences
Proteine und Peptide
Cathepsin inhibitor
Irreversible, non-toxic, cell-permeable inhibitor of cysteine proteases calpain, cathepsins B and L, and Trypanosome trypanopain. Can be used in vitro (typically 20 µM) or in vivo. Inhibits tumor formation and growth.