Clostridium difficile toxin B
Lieferant: ENZO LIFE SCIENCES
BMLG1500050
ENZOBMLG1500050EA
1190
EUR
ENZOBMLG1500050
Clostridium difficile toxin B
Mikrobiologische Testsysteme
Antigen-Antikörper-Tests
Ag-AK, Seren
Clostridium difficile toxin B from Clostridium difficile, strain VPI10463 is a high purity toxin applicable to ELISA, antibody capture, cytotoxicity studies and LINE assay.
- High potency retained following inactivation
- Minimal lot-to-lot variation
- Lyophilised for ease of handling and long term stability