CTT, Enzo Life Sciences
Lieferant: ENZO LIFE SCIENCES
BMLPI1360001
ENZOBMLPI1360001EA
161
EUR
ENZOBMLPI1360001
CTT, Enzo Life Sciences
Proteine und Peptide
MMP inhibitor
CTTHWGFTLC is a specific inhibitor of the gelatinases, MMP-2 and MMP-9, with an IC50 of 10-50 µM. Can be used in in vitro studies, zymography and in situ zymography, in tissue culture, and in vivo. Also available, negative control peptide STTHWGFTLS.