In-situ hybridisation buffer (1.25X concentrate), Enzo Life Sciences
Lieferant: ENZO LIFE SCIENCES
ENZ33808
ENZOENZ33808EA 88 EUR
ENZOENZ33808
In-situ hybridisation buffer (1.25X concentrate), Enzo Life Sciences
Puffer Elektrophorese-Puffer Hybridisierungspuffer
In Situ Hybridization Buffer provides a high quality hybridization medium for in situ hybridization analyses using either DNA or oligonucleotide probes labeled with biotin-, digoxigenin- or fluorescein-modified nucleotides.
Bestellinformation: Quantity: Provides a sufficient amount of hybridization buffer concentrate to prepare 12.5ml of ready-to-use probe reagent solutions.