In-situ hybridisation buffer for HPV probes
Lieferant: ENZO LIFE SCIENCES
ENZ339050050
ENZOENZ339050050EA 234 EUR
ENZOENZ339050050
In-situ hybridisation buffer for HPV probes
Puffer Elektrophorese-Puffer Hybridisierungspuffer
In situ hybridisation buffer for HPV probes (ready-to use) provides a high quality hybridisation medium for In situ hybridisation analyses using either DNA or oligonucleotide probes labeled with biotin-, or fluorescein-modified nucleotides.
  • Optimised buffer to use with HPV collection of probes