Cyanine 5-UTP (enhanced)
Lieferant: ENZO LIFE SCIENCES
ENZ42506
ENZOENZ42506EA
467
EUR
ENZOENZ42506
Cyanine 5-UTP (enhanced)
Nukleinsäure-Reagenzien
Nukleotide
Cyanine 5-UTP (Enhanced) can replace UTP as a substrate for T7 RNA polymerase in labeling systems that generate labeled probes through in vitro transcription. Probes generated by these methods are suitable for multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with cyanine 3-UTP.