Roter Langzeit-Zelltracer, CYTO-ID®
Lieferant: ENZO LIFE SCIENCES
CYTO-ID® is a live cell fluorescent labelling used for a long term cell viability, cytotoxicity, cell adhesion, cell migration, and cell-cell fusion studies.
- Dual labeling with CELLESTIAL® fluorescent probes
- Minimal transfer of fluorescence from dye-labeled to unlabelled cells
Uses proprietary non covalent cell labeling technology to stably incorporate a red fluorescent dye containing hydrophobic aliphatic chains into the cell membrane's lipid bilayer.
Bestellinformation: Beinhaltet einen roten Tracer-Farbstoff (50 µl), 4X Markierungspuffer (12,5 ml) und 10X HBSS (25 ml).
Lieferumfang: Kit contains CYTO-ID® red tracer dye, 50 µl, 4X labeling buffer, 12,5 ml and 10X HBSS, 25 ml.