Nuclear-ID™ Blue/Green cell viability reagent
Lieferant: ENZO LIFE SCIENCES
ENZ53004C100
ENZOENZ53004C100EA 368 EUR
ENZOENZ53004C100
Nuclear-ID™ Blue/Green cell viability reagent
Proteinkonjugate und Markierungen
Nucleus dye
The Nuclear-ID™ Blue/Green cell viability reagent (Prod. No. ENZ-53004) is a mixture of a blue fluorescent cell-permeable nucleic acid dye and a green fluorescent cell-impermeable nucleic acid dye that is suited for staining dead nuclei. The staining pattern arising from the simultaneous combination of these two dyes permits determination of live and dead cell populations by fluorescence/confocal microscopy. The reagent, supplied as a 1000x solution, is sufficient for 1000 microscopy assays. The single-tube format makes this cell viability reagent easy to use. It leaves the cytoplasm unstained, potentially allowing visualization of other cell markers.
Bestellinformation: Quantity: 100 μl (for 1000 microscopy assays).