T7 RNA polymerase
Lieferant: Thermo Fisher Scientific
T7 RNA polymerase is ideal for synthesis of unlabeled and labeled RNA. It catalyses the 5'→3' synthesis of RNA on either single-stranded DNA or double-stranded DNA downstream from it promoter. It is also used in the study of RNA secondary structure and RNA-protein interactions, RNA splicing.