RevertAid™ Reverse Transcriptase
Lieferant: Thermo Fisher Scientific
EP0441
EP0442
EP0441EA
91.5
EUR
EP0441
EP0442
RevertAid™ Reverse Transcriptase
Nukleinsäure-Reagenzien
RT-PCR und cDNA Reagenzien
RevertAid™ Reverse Transcriptase (RT) is a genetically modified M-MuLV RT. It differs from the M-MuLV RT by its structure and catalytic properties. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity and a RNase H activity specific to RNA in RNA-DNA hybrids, which is significantly lower than that of Avian Myeloblastosis Virus (AMV) reverse transcriptase.
- Efficient synthesis of full length first strand cDNA up to 13 kb
- Optimum activity at 42 °C
- Active up to 50 °C
- Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, fluorescein-labelled nucleotides)
Lieferumfang: Supplied with 5X reaction buffer.