Eppendorf® Tube Racks
Lieferant: EPPENDORF
0030119.800 0030119.819 0030119.827 0030119.843 0030119.851 0030119.835 0030119.860
EPPE0030119.800EA 51.5 EUR
EPPE0030119.800 EPPE0030119.819 EPPE0030119.827 EPPE0030119.843 EPPE0030119.851 EPPE0030119.835 EPPE0030119.860
Eppendorf® Tube Racks
Gestelle Reagenzglasgestelle
Optimize workflows through user-friendly handling of clearly arranged samples. Eppendorf® Tube Racks are ideal for sample processing, transport, and short- to mid-term storage of samples.
  • Stackable within one rack format and also between all rack variants; saves laboratory space when not in use
  • Non-slip silicone feet and a tilt-proof footprint design ensure safe handling
  • Made of polypropylene (PP) for high chemical and physical resistance
  • High flexibility; different sized tubes fit in the 1.5/2.0 ml, 5.0/15.0 ml, 5.0/15/50 ml, and cryogenic tube rack
More Product Information