PCR plates, Eppendorf twin.tec® LoBind
Lieferant: EPPENDORF
0030129.571
EPPE0030129.571EA 1855 EUR
EPPE0030129.571
PCR plates, Eppendorf twin.tec® LoBind
Mikrotiterplatten PCR-Platten
Eppendorf twin.tec® PCR plates combine extremely thin walled polypropylene wells for optimal heat transfer to the sample and robust polycarbonate frames for ultimate rigidity and torque-resistance. Eppendorf twin.tec® plates reduce the risk of cross contamination with raised well rims for effective sealing.
  • PCR clean
These Eppendorf twin.tec® 96 plates are ideal for quantitative Real-Time PCR, standard PCR, high-throughput PCR and small volume sample handling.
Bestellinformation: PCR plates are available with barcode upon request, please contact your local Avantor sales office for further details.