Pipette holder 2 and charger shell 2 for stands and carousels
Lieferant: EPPENDORF
3116000.112 3116000.120 3116000.139 3116000.147 3116602.007 3116603.003
613-6972EA 34.9 EUR
613-6972 EPPE3116000.120 EPPE3116000.139 EPPE3116000.147 EPPE3116602.007 EPPE3116603.003
Pipette holder 2 and charger shell 2 for stands and carousels
Pipettenständer
A broad range of holders for all current manual and electronic Eppendorf liquid handling instruments and most predecessors.
Place your pipette and multipette where you need them: Mounted to a wall, on the shelf above your bench or inside a biological safety cabinet. In addition, all holders can be inserted into pipette carousel 2. Charger shells for installation into charger carousel 2 are available as well.