epMotion® 96 Flex Liquid Handling System
Lieferant: EPPENDORF
5069000.519
EPPE5069000.519EA 12550 EUR
EPPE5069000.519
epMotion® 96 Flex Liquid Handling System
Automatisierte Liquid-Handling-Systeme
Elevate lab workflows with the epMotion™ 96 Flex. Designed for simultaneous 96-channel pipetting for convenient processing of both 96-well and 384-well plates.
  • System covers 0,5 to 1000 µl with only two interchangeable dispensing heads
  • Ergonomic handling, even in clean benches
  • Base-mounted touchscreen user interface with intuitive software
  • High precision according to ISO 23783 standards
  • Suitable for use in laminar flow cabinets
  • Only compatible with epT.I.P.S.™ Motion Reloads
The epMotion® 96 Flex revolutionises liquid handling by providing speed and accuracy for any laboratory environment. The intuitive design supports seamless integration into biosafety cabinets, facilitating ergonomic and efficient workflows. With exchangeable dispensing heads, it accommodates a wide volume range, ensuring flexibility for various applications such as plate reformatting, magnetic bead clean-up, and immunoassays. Certified according to ISO 23783-2 standards, it guarantees exceptional precision and reproducibility, making assays more reliable. The compact design and user-friendly interface simplify operations, significantly reducing processing time compared to traditional pipettes.
The epMotion® 96 Flex base device is compatible with 96-well and 384-well microplates, PCR, and deepwell plates. The unit features a wide variety of programmable modes, including pipette, multidispense, multiaspirate, pipette + mix, manual pipette, dilute + mix, reverse pipetting, small volume, programming and set height.
The epMotion® 96 Flex’s exchangeable dispensing heads accommodate a very large volume range and can handle less than 96 tips. This design provides full flexibility for current and future applications, with the possibility to use two tip sizes per dispensing head to maximise accuracy and precision. Dispensing heads are easily exchanged, with no tools required.
Standards:
The epMotion® 96 Flex base device and dispensing heads are certified according to ISO 23783-2 standards.
Zubehörinformationen:
The accessory TipHolder is designed for use with epT.I.P.S™ Motion Reloads. The TipHolder is reusable and autoclavable.
Bestellinformation:
Base device and dispensing heads are sold separately. Dispensing heads, power cable, and accessories must be ordered separately. One (1) TipHolder is included with each epMotion® 96 Flex base device. Dispensing heads are supplied with hard shell case for transport and storage.