MixMate® Mixer
Lieferant: EPPENDORF
5353000.510
EPPE5353000.510EA 2120 EUR
EPPE5353000.510
MixMate® Mixer
Schüttler Mikrotiterplatten-Schüttler
Mix samples completely and reliably with the Eppendorf® MixMate. 96 and 384-well plates and reaction tubes are mixed by the 2DMix-Control technology even at the microliter scale. The additional vortex function has a 2 s run timer allowing processing your next sample without restarting.
  • High-speed mixer (3000 rpm) for fast and efficient mixing of small volumes in all common vessel formats up to 50 ml
  • 2DMix-Control technology (planar mixing orbit without vertical movement) for controlled mixing and reproducible results in seconds
  • Anti-spill technology for controlled mixing movements without lid wetting and cross-contamination
  • Five different tube holders (0,5 ml, 1,5/2,0 ml, 5/15 ml, 30/50 ml, PCR 96-well plates) for high vessel flexibility
  • Integrated ergonomic vortex function for various vessel formats for distinct sample mixing
  • Minimal hand/arm vibration transfer during vortexing (certificate available) for ergonomic operation (PhysioCare Concept®)
  • Five soft keys with pre-defined standard mixing parameters for easy usage
  • Very stable remains stationary even at maximum mixing speed
  • Whisper quiet operation to benefit the work environment
Mix samples in seconds, fully and reliably. Whether in plates (96 or 384-wells) or reaction tubes, your samples will be optimally processed. With the integrated ergonomic vortex function, the MixMate is your perfect laboratory assistant.
Other functions include 2D Mix-control, anti-spill technology, touch vortexing.
Mixing and vortexing radius: 1,5 mm