Thermal cyclers, Mastercycler® nexus flat
Lieferant: EPPENDORF
6330000.013
EPPE6330000.013EA 0 EUR
EPPE6330000.013
Thermal cyclers, Mastercycler® nexus flat
PCR-Cycler
Special applications require special consumables. With its flat block and no wells, the Mastercycler® nexus flat offers the optimal foundation for slides and other unconventional consumable formats. When doing in situ PCR, results are normally influenced by the characteristic temperature transfer of the corresponding in situ adapter. The Mastercycler® nexus flat can heat and cool slides directly without the need for an adapter.
  • Flat block without wells
  • Intuitive graphic programming
  • Small footprint
  • Up to two more units can be connected to the central unit
  • E-mail notification
  • Optional self-test functionality
*A Mastercycler® nexus without 'eco' suffix is needed to run a Mastercycler® nexus flat eco. Up to 2 Mastercycler® nexus flat eco can be controlled by one Mastercycler® nexus without 'eco' suffix.