ePrep µSPEed® Cartridge Rack
Lieferant: EPREP
01-04161
EPRE01-04161EA 1980 EUR
EPRE01-04161
ePrep µSPEed® Cartridge Rack
Filter Filterkartuschen
The µSPEed Cartridge rack is available where the use of µSPEed cartridge is required. The rack can hold up to 96 cartridges and can be fully integrated with ePrep Workflow.
Bestellinformation: Cartridges sold separately.