Sample bottles, narrow neck, round, HDPE, Boston type
Lieferant: EP SCIENTIFIC
319-1000 319-0125
EPSC319-1000EA 151 EUR
EPSC319-1000 EPSC319-0125
Sample bottles, narrow neck, round, HDPE, Boston type
Flaschen Kunststoffflaschen
HDPE container with white LDPE-lined polypropylene cap.
  • Narrow mouth for precision pouring
  • Caps assembled onto containers
  • Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride
  • Certificate of Analysis (CofA) included in the case and barcoded with lot number for traceability
  • Each container individually barcoded with lot number and unique container number
  • Analysis labels included unattached for optional use
  • Carton sealed with security tape providing tamper evidence
Recommended for collecting water samples and otherMore Product Information