Restriction enzymes, BamHI, HC, Fermentas
Lieferant: Thermo Fisher Scientific
ER0053
ER0053EA
144
EUR
ER0053
Restriction enzymes, BamHI, HC, Fermentas
Nukleinsäure-Reagenzien
Restriktionsenzym
BamHI restriction enzyme recognises G^GATCC sites and cuts best at 37 °C in its own unique buffer.
- Superior quality, stringent quality control and industry leading manufacturing process
- Convenient colour-coded five buffer System
- Includes universal Tango buffer for double-digestions
- BSA premixed in reaction buffers