Signs, duraview, DuraStripe®
Lieferant: ERGOMAT
DVA421W-VERT
ERGODVA421W-VERTEA 109 EUR
ERGODVA421W-VERT
Signs, duraview, DuraStripe®
Warn- und Verbotszeichen
DuraView's attention grabbing yellow border protects US letter and A4 sized paper inserts on three sides for easy exchange flexibility.
Choose to keep the long side or short side of the DuraView open for horizontal or vertical installation orientation.
DuraView is perfect for temporary or permanent floor signs, safety warnings, lean instructions, location indicators, bar codes, or any other information you would use signage to convey.
Sizes come in A4 (210,82×297,8 mm).