Precision scale magnifiers
Lieferant: ESCHENBACH OPTIK
155074
ESCH155074EA 177 EUR
ESCH155074
Precision scale magnifiers
Lupen
Precision magnifiers with brightness control and removable measuring scale available with battery or mains handle with brightness control.
Aspheric PXM® lightweight lens with Cera-tec® hard coating (both sides).