Reading magnifier
Lieferant: ESCHENBACH OPTIK
2668950
ESCH2668950EA 96.2 EUR
ESCH2668950
Reading magnifier
Lupen
This aplanatic magnifier is suitable for reading and looking at pictures for long periods.
  • Image free from spherical aberration
  • Edge-to-edge and distortion-free image
  • Comfortable thanks to the ergonomic handle