Temperature and humidity data loggers, iMINI
Lieferant: Askey Dataloggers
MX-OE-S-8-L
ESCOMX-OE-S-8-LEA 153 EUR
ESCOMX-OE-S-8-L
Temperature and humidity data loggers, iMINI
Datenlogger
The iMINI data loggers are designed for the monitoring of both temperature and humidity on sensitive products. They are available as disposable or reusable loggers. They encompass a full spectrum of uses from food processing and warehousing, to blood storage, transport and clinical trials.
  • Visual alarms: Two red and two blue LED
  • Large LCD display with start and stop buttons
  • NIST Traceable® calibration and bookmarking capability
Standards: ISO 9001:2008, CE, RoHS compliant; accuracy certified.