Abbe refractometer
Lieferant: EUROMEX
98.490
EUMX98.490EA 0 EUR
EUMX98.490
Abbe refractometer
Refraktometer Tischrefraktometer
The Abbe laboratory refractometer is a bench-top instrument for high-precision measurements of an index of refraction.
  • More accurate than hand refractometers
  • Capable of measuring all kinds of concentrations and identifying several types of substances
It can be connected to a water bath for measuring at a controlled temperature and used for determining the refractive index of glass, plastic films. During operating this type of refractometer, the use of a cold light source is recommended. The instrument is equipped with a built-in thermometer and water connection to control fluid temperatures.
This Abbe refractometer has scales for measurement of sugar concentrations and refractive indices.
The use of a cold light source like the LE.5209 is recommended for this type of refractometer.
Lieferumfang: Supplied with flight case, thermometer, test plate and adjusting tools.