High Speed Camera, CMEX-5 Pro
Lieferant: EUROMEX
DC.5000-PRO
EUMXDC.5000-PROEA 775 EUR
EUMXDC.5000-PRO
High Speed Camera, CMEX-5 Pro
Kameras
High speed camera with perfect colour-reproduction capability.
  • 5.1 Mpix CMOS color USB 3.0 camera
  • 1/2,5" sensor, 2560×1922 pixels
  • Low signal/noise ratio
  • USB 3.0 high speed interface (USB 2.0 compatible)
Housing: Integrated CNC aluminum alloy
Power supply: 5 V over PC USB port
Lieferumfang: Supplied with 0,5× objective with C-mount, USB 3 cable, 30 and 30,5 mm adapters for stereomicroscopes, 76×24 mm calibration slide (1 mm/100), carton box.