5-(Ethylthio)-1H-tetrazol (ETT) 0.25 mol/l in Acetonitril
Lieferant: EUROGENTEC
30-3140-52
30-3140-45
30-3140-52
30-3140-57
30-3140-61
30-3140-62
EURG30-3140-52EA
0
EUR
EURG30-3140-52
GLEN30-3140-45
GLEN30-3140-52
GLEN30-3140-57
GLEN30-3140-61
GLEN30-3140-62
5-(Ethylthio)-1H-tetrazol (ETT) 0.25 mol/l in Acetonitril
5-(Ethylthio)-1H-tetrazol (ETT)
Formel:
C₃H₆N₄S |
MDL:
MFCD00068733 CAS-Nummer: 89797-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...