pH electrode with spear tip, Eutech

Lieferant: EUTECH
EC620133
EUTEEC620133EA 369 EUR
EUTEEC620133
pH electrode with spear tip, Eutech
Elektroden pH-Elektroden
This glass-body electrode features a spear tip. Suitable for semi-solid samples.

  • Open pore junction
  • For use with all pH meters with BNC input connector
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR