Dissolved oxygen electrode, Eutech
Lieferant: EUTECH
ECDOHANDYNEW
EUTEECDOHANDYNEWEA 874 EUR
EUTEECDOHANDYNEW
Dissolved oxygen electrode, Eutech
Elektroden Sauerstoff-Elektroden
This galvanic DO electrode with ATC has a response time of 40 sec to reach 93 % of the final reading. For use with the following Eutech meters: DO 600 / DO 300 / DO 110 / PD 650 / PCD 650.
Minimum sample flow: 2 inch / sec
Maximum pressure: 7,5 bar
Lieferumfang: Supplied with one assembled membrane cap housing, one refilling electrolyte and one scouring pad.