ORP electrode, Eutech
Lieferant: EUTECH
ECFC7960101B
EUTEECFC7960101BEA 201 EUR
EUTEECFC7960101B
ORP electrode, Eutech
Elektroden Redox-Elektroden
General purpose plastic-body ORP electrode for use with all pH meters with BNC input connector.