Multi-parameter meters, handheld, Eutech CyberScan PCD 650
Lieferant: EUTECH
ECPCDWP65044K
EUTEECPCDWP65044KEA 3150 EUR
EUTEECPCDWP65044K
Multi-parameter meters, handheld, Eutech CyberScan PCD 650
Elektrochemische Messgeräte pH- und Multi-Parameter-Messgeräte
This portable meter enables users to measure pH, mV, conductivity, TDS, salinity, ions, dissolved oxygen and temperature values. It simultaneously measures and displays up to four parameters.
  • Accepts two-cell and four-cell conductivity probe to measure up to 500 m/s — meter measures pure water
  • Selectable multi-point calibration for more accurate conductivity measurements or single-point calibration for quick, convenient calibration with no mistakes
  • Electrode status report, calibration data and simultaneous display of up to 4 parameters
  • Rugged and IP67-rated waterproof for applications in harsh environments; meter floats on water
  • Recorded readings individually stamped with time and date in GLP compliant format
  • HI-LO limit alarm for easy quality control – Meter alerts when readings fall outside selected ranges
  • Wireless Information Technology: No wires, no cables — Transfer data from meter to computer with the press of a button
Ideal for environmental testing, water quality testing, industrial applications and aquaculture.
Lieferumfang: Supplied with pH electrode ECFC7252203B, 4-cell conductivity electrode ECCONSEN9203J, dissolved oxygen probe ECDOHANDYNEW (all electrodes come with 3 m cables), DAS software, multi-probe holder, power adapter and carrying kit set incl. a plastic carrying case, 1× pH 4.01, pH 7.00 buffer solutions, storage solution and 1× deionised (rinse) water (60 ml each).