ATC probes, Eutech
Lieferant: EUTECH
ECPH5TEM01P ECPHWPTEM01W
EUTEECPH5TEM01PEA 65.5 EUR
EUTEECPH5TEM01P EUTEECPHWPTEM01W
ATC probes, Eutech
ATC electrodes for use with Eutech pH 5+, pH 6+ and ION 6+ meters or for use with CyberScan pH 300 and 310 meters.