Anti-PAX8 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB05513
EVBIEB05513EA 339 EUR
EVBIEB05513
Anti-PAX8 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PAX8 (paired box 8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR