Anti-SF3B3 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB05543
EVBIEB05543EA 339 EUR
EVBIEB05543
Anti-SF3B3 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SF3B3 (splicing factor 3b, subunit 3, 130kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR