Anti-ARL4A Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB05549
EVBIEB05549EA 339 EUR
EVBIEB05549
Anti-ARL4A Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARL4A (ADP-ribosylation factor-like 4A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR