Anti-NFATC4 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB05562
EVBIEB05562EA 339 EUR
EVBIEB05562
Anti-NFATC4 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: NFATC4 (nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR