Anti-FHL3 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB06509
EVBIEB06509EA 339 EUR
EVBIEB06509
Anti-FHL3 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: FHL3 (four and a half LIM domains 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR