Anti-VPS29 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB07369
EVBIEB07369EA 339 EUR
EVBIEB07369
Anti-VPS29 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VPS29 (vacuolar protein sorting 29 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR