Anti-APBB1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB07373
EVBIEB07373EA 339 EUR
EVBIEB07373
Anti-APBB1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: APBB1 (amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR