Anti-SEPT1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB07412
EVBIEB07412EA 339 EUR
EVBIEB07412
Anti-SEPT1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SEPT1 (septin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR