Anti-PENK Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08195
EVBIEB08195EA 339 EUR
EVBIEB08195
Anti-PENK Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PENK (proenkephalin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR