Anti-APOBEC2 Goat polyclonal antibody unconjugated

Lieferant: Everest Biotech
EB08444
EVBIEB08444EA 339 EUR
EVBIEB08444
Anti-APOBEC2 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: APOBEC2 (apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR