Anti-TGFBR3 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08467
EVBIEB08467EA 339 EUR
EVBIEB08467
Anti-TGFBR3 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TGFBR3 (transforming growth factor, beta receptor III)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR