Anti-ARSD Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08469
EVBIEB08469EA 339 EUR
EVBIEB08469
Anti-ARSD Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ARSD (arylsulfatase D)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR