Anti-NUAK1 Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08474
EVBIEB08474EA 335 EUR
EVBIEB08474
Anti-NUAK1 Goat Polyclonal Antibody
Type: Primary
Antigen: NUAK1 (NUAK family, SNF1-like kinase, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR