Anti-FTO Goat Polyclonal Antibody

Lieferant: Everest Biotech
EB08564
EVBIEB08564EA 335 EUR
EVBIEB08564
Anti-FTO Goat Polyclonal Antibody
Type: Primary
Antigen: FTO (fat mass and obesity associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Dog, Human, Mouse, Pig, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR